Learn More
Microsolv Technology Corporation Caps, Snap Top, with White Silicone Rubber / Tan PTFE Septa, fitted in smooth, polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502270281
Encompass_Preferred
Caps, Snap Top, with White Silicone Rubber / Tan PTFE Septa, fitted in smooth, polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
This item is not returnable. View return policy
Catalog No. 50-227-0281 Supplier Microsolv Technology Corporation Supplier No. 9502C11MB
Caps, Snap Top, with White Silicone Rubber / Tan PTFE Septa, fitted in smooth, polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.