Learn More
Microsolv Technology Corporation Caps, Snap Top, with Silicone Rubber / PTFE Pre-Slit (Y Shape) Septa, fitted in smooth, polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502271242
Encompass_Preferred
Pre-Slit septa are normally recommended for Waters Alliance instruments.Y Shape Pre-Slit offers a great target area for injector needles and are easier to penetrate but do not reseal as well as single slit septa.
This item is not returnable. View return policy
Catalog No. 50-227-1242 Supplier Microsolv Technology Corporation Supplier No. 9502C34CB
Pre-Slit septa are normally recommended for Waters Alliance instruments.Y Shape Pre-Slit offers a great target area for injector needles and are easier to penetrate but do not reseal as well as single slit septa.