Learn More
Microsolv Technology Corporation Caps, Snap Top, with White PTFE Septa, fitted in smooth, polyethylene, Green Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 1000/CS.
Supplier:  Microsolv Technology Corporation 9502C50CGM
Encompass_Preferred
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal.
Catalog No. 50-227-1257