Learn More
Microsolv Technology Corporation Caps, Snap Top, with White PTFE Septa, fitted in smooth, polyethylene, Green Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502271256
Encompass_Preferred
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal.
This item is not returnable. View return policy
Catalog No. 50-227-1256 Supplier Microsolv Technology Corporation Supplier No. 9502C50CG
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal.