Learn More
Microsolv Technology Corporation Caps, Snap Top, with White PTFE Septa, fitted in smooth, polyethylene, Red Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 1000/CS.
Catalog No. 502271259
Encompass_Preferred
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal.
This item is not returnable. View return policy
Catalog No. 50-227-1259 Supplier Microsolv Technology Corporation Supplier No. 9502C50CRM
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal.