Learn More
Microsolv Technology Corporation Caps, Snap Top, with White PTFE Septa, fitted in smooth, polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 1000/CS.
Catalog No. 502271255
Encompass_Preferred
This PTFE (only) Septa is for single injections or non volatile samples as it will not reseal. Buying in case quantities can save money and time.
This item is not returnable. View return policy
Catalog No. 50-227-1255 Supplier Microsolv Technology Corporation Supplier No. 9502C50CBM
This PTFE (only) Septa is for single injections or non volatile samples as it will not reseal. Buying in case quantities can save money and time.