Learn More
Microsolv Technology Corporation Caps Snptp Blue Soft Pe (X)
Catalog No. 5001452036
Encompass_Preferred
Caps, Snap Top, with 1.00mm thick, White Silicone Rubber / Blue PTFE Pre-Slit (X Shape) Septa, fitted in smooth, soft polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52036 Supplier Microsolv Technology Corporation Supplier No. 9502CS30B
Caps, Snap Top, with 1.00mm thick, White Silicone Rubber / Blue PTFE Pre-Slit (X Shape) Septa, fitted in smooth, soft polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. MicroSolv Brand. 100/PK.