Learn More
Microsolv Technology Corporation Caps Solid White Tfe/Sil
Catalog No. 5001452227
Encompass_Preferred
Caps, Screw Top, Solid, with a Silicone Rubber / PTFE liner, in knurled, polypropylene, White Caps. For use with 13-425mm thread,15x45mm autosampler vials. MicroSolv Brand 100/PK.
Catalog No. 50-014-52227 Supplier Microsolv Technology Corporation Supplier No. 9504S05SW
Caps, Screw Top, Solid, with a Silicone Rubber / PTFE liner, in knurled, polypropylene, White Caps. For use with 13-425mm thread,15x45mm autosampler vials. MicroSolv Brand 100/PK.