Learn More
Microsolv Technology Corporation Caps Storage 18-400 Thread Red
Catalog No. 5001452429
Encompass_Preferred
Caps, Screw Top, Solid, with Red Butyl Rubber / Grey PTFE Liner, fitted in knurled polypropylene, Black Caps. For use with ND18 thread, storage Vials or Bottles. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52429 Supplier Microsolv Technology Corporation Supplier No. 97018SP
Caps, Screw Top, Solid, with Red Butyl Rubber / Grey PTFE Liner, fitted in knurled polypropylene, Black Caps. For use with ND18 thread, storage Vials or Bottles. MicroSolv Brand. 100/PK.