Learn More
Microsolv Technology Corporation Caps Storage Vials Sm Pe Snapt
Catalog No. 5001452418
Encompass_Preferred
Caps, Push Top, Solid, Translucent Polyethylene. No liner required with ND18 Push Top, Econo-Storage Vials. BASIK Brand. 100/PK.
Catalog No. 50-014-52418 Supplier Microsolv Technology Corporation Supplier No. 97010SC18
Caps, Push Top, Solid, Translucent Polyethylene. No liner required with ND18 Push Top, Econo-Storage Vials. BASIK Brand. 100/PK.