Learn More
Electron Microscopy Sciences Caps for Syringe
SDP
Catalog No. 50949293
Encompass_Preferred
One size fits-all syringe caps that can be used with either a luer slip or luer locking syringe tip. These tips make the syringes suitable for usage as embedding media storage vessels. 
Catalog No. 50-949-293 Supplier Electron Microscopy Sciences Supplier No. 7250005
One size fits-all syringe caps that can be used with either a luer slip or luer locking syringe tip. These tips make the syringes suitable for usage as embedding media storage vessels. 