Learn More
Bioworld CAPS, Transfer Buffer (5X), 500 mL
Catalog No. 50198637
Encompass_Preferred
Product DescriptionCAPS-based transfer buffer at 5X contains a concentration between 90-110 mM CAPS and 10% methanol, with a pH value between 10.8-11.2 at 25C°. Buffer may be preferable for transfers of high molecular weight proteins (such as >150 kD) and in cases where the glycine component in buffer may interfere with downstream protein sequencing applications. Protease (P/F): None
Catalog No. 50-198-637 Supplier Bioworld Supplier No. 214200161
Product DescriptionCAPS-based transfer buffer at 5X contains a concentration between 90-110 mM CAPS and 10% methanol, with a pH value between 10.8-11.2 at 25C°. Buffer may be preferable for transfers of high molecular weight proteins (such as >150 kD) and in cases where the glycine component in buffer may interfere with downstream protein sequencing applications. Protease (P/F): None