Learn More
Chemglass Life Sciences 20-400 Replacement Cap with Opening, Minum-Ware
SDP
Catalog No. 501532672
Encompass_Preferred
20-400 Replacement Cap with Opening, Minum-Ware. Used with Viton O-Rings and threaded outer joints for making grease-free, vacuum tight seals on 14/10 MINUM-WARE glassware components. The 8-425 size is used on the sidearm of Hickman still head and MW-81 GC collection vial, while the 13-425 is used on the MW-56 two neck flask sidearm.
Catalog No. 50-153-2672 Supplier Chemglass Life Sciences Supplier No. MW10233
20-400 Replacement Cap with Opening, Minum-Ware. Used with Viton O-Rings and threaded outer joints for making grease-free, vacuum tight seals on 14/10 MINUM-WARE glassware components. The 8-425 size is used on the sidearm of Hickman still head and MW-81 GC collection vial, while the 13-425 is used on the MW-56 two neck flask sidearm.