Learn More
Cayman Chemical Capsacn 100mg
SDP
Supplier:  Cayman Chemical 92350500 MG
Encompass_Preferred
A terpene alkaloid; induces inward currents in HEK293 cells expressing rat TRPV1 (EC50 = 0.64 µM at neutral pH); decreases LPS-induced PGE2 production, as well as reduces LPS- and IFN-induced NO release in isolated mouse peritoneal macrophages at 10 and 50 µM; induces substance P release in rat spinal cord slices (EC50 = 2.3 µM); reduces acetylcholine- or phenylquinone-induced writhing (ED50s = 1.33 and 1.38 mg/kg, respectively, s.c.) but has no effect on the latency to paw withdrawal in the hot plate test in mice (ED50 = >20 mg/kg, s.c.)
Catalog No. 50-289-4721