Learn More
Enzo Life Sciences Capsaicin (200mg). CAS: 404-86-4
SDP
Supplier:  Enzo Life Sciences BMLEI1250200
Encompass_Preferred
Capsaicin is an agonist at the TRPV1 receptor, a ligand-gated non-selective cation channel. Exhibits neuroprotective effects in a model of transient global cerebral ischemia. Induces release of substance P from afferent nociceptive specific neurons. Constituent of cayenne pepper. Powerful excitant of peropheral sensory nerve endings, specifically unmyelinated afferent neurons (C-fibers). Reversibly inhibits aggregation of platelets. Alternative name: 8-Methyl-N-vanillyl-6-nonenamide. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol, methanol or chloroform. Sparingly soluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-0562