Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capsaicin beta-D-Glucopyranoside 90.0+%, TCI America™
SDP

Catalog No. C2548200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2548200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2548200MG Supplier TCI America Supplier No. C2548200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Non-pungent capsaicin derivative for laboratory research use.

Chemical Identifiers

CAS 153409-16-6
Molecular Formula C24H37NO8
Molecular Weight (g/mol) 467.559
InChI Key HEYWYCJIIXVRPS-SOFGYWHQSA-N
PubChem CID 56973704
IUPAC Name (E)-N-[[3-methoxy-4-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]methyl]-8-methylnon-6-enamide
SMILES CC(C)C=CCCCCC(=O)NCC1=CC(=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O)OC

Specifications

Color White
Quantity 200 mg
Formula Weight 467.56
Percent Purity ≥90.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Capsaicin beta-D-Glucopyranoside
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.