Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Capsaicin β-D-glucopyranoside | 153409-16-6 | 90.55% | 467.55 | 10 MG
SDP

Catalog No. 5000425550
Encompass_Preferred

Capsaicin β-D-glucopyranoside is a glucoside converted by Capsaicin. Capsaicin is the active ingredient of chili peppers and gives them the characteristic pungent flavor.

  • Solid appearance
  • White to off-white color
  • Molecular formula: C24H37NO8
  • Derived from plants, solanaceae, capsicum annuum L.
  • Storage as solid at -20°C, sealed, away from moisture and light
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-25550 Supplier Medchemexpress LLC Supplier No. HYN945210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Capsaicin β-D-glucopyranoside is a glucoside converted by Capsaicin. Capsaicin is the active ingredient of chili peppers and gives them the characteristic pungent flavor.

  • Solid appearance
  • White to off-white color
  • Molecular formula: C24H37NO8
  • Derived from plants, solanaceae, capsicum annuum L.
  • Storage as solid at -20°C, sealed, away from moisture and light
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.