Learn More
Cayman Chemical CapsaIcIn-d3 1mg
SDP
Catalog No. 502910271
Encompass_Preferred
An internal standard for the quantification of capsaicin by GC- or LC-MS
Catalog No. 50-291-0271 Supplier Cayman Chemical Supplier No. 346951 MG
An internal standard for the quantification of capsaicin by GC- or LC-MS