Learn More
Selleck Chemical LLC Capsaicin(Vanilloid) 50mg 404-86-4 Qutenza, Vanilloid
SDP
Supplier:  Selleck Chemical LLC S199050MG
Encompass_Preferred
Capsaicin (Qutenza, Vanilloid) is an active component of chili peppers, which are plants belonging to the genus Capsicum.?Capsaici is a TRPV1 agonist with an EC50 of 0.29 ?M in HEK293 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8224