Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capsanthin (=Paprika Extract) (Vegetable oil solution), TCI America™
SDP

Catalog No. C078125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C078125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C078125G Supplier TCI America Supplier No. C078125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 465-42-9
Molecular Formula C40H56O3
Molecular Weight (g/mol) 584.89
MDL Number MFCD03423725
InChI Key VYIRVAXUEZSDNC-RDJLEWNRSA-N
Synonym Paprika Extract
PubChem CID 5281228
ChEBI CHEBI:3375
IUPAC Name (2E,4E,6E,8E,10E,12E,14E,16E,18E)-1-[(1R,4S)-4-hydroxy-1,2,2-trimethylcyclopentyl]-19-[(4R)-4-hydroxy-2,6,6-trimethylcyclohex-1-en-1-yl]-4,8,13,17-tetramethylnonadeca-2,4,6,8,10,12,14,16,18-nonaen-1-one
SMILES C\C(\C=C\C=C(/C)\C=C\C(=O)[C@]1(C)C[C@@H](O)CC1(C)C)=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C[C@@H](O)CC1(C)C

Specifications

Color Red
Quantity 25 g
Formula Weight 584.89
Physical Form Liquid
Chemical Name or Material Capsanthin (=Paprika Extract) (Vegetable oil solution)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.