Learn More
Enzo Life Sciences Capsazepine (5mg). CAS: 138977-28-3
SDP
Supplier:  Enzo Life Sciences ALX550145M005
Encompass_Preferred
Analog of capsaicin (BML-EI125-0200) that acts as a capsaicin antagonist. Inhibits resiniferatoxin and capsaicin-induced contractions of guinea pig trachea. Inhibits C-fiber-smooth muscle contractions induced by capsaicin, bradykinin and neurokinin A. Increases intracellular Ca2+ levels. Alternative name: N-[2-(4-Chlorophenyl)ethyl]-1,3,4,5-tetrahydro-7,8-dihydroxy-2H-2-benzazepine-2-carbothioamide. Purity: ≥97% (HPLC, TLC). Appearance: White to off-white powder. Solubility: Soluble in methanol, 100% ethanol (25mg/ml) or DMSO (25mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-9489