Learn More
Cytiva 1K Minimate™ Tangential Flow Filtration (TFF) capsule with Omega™ membrane (1/pkg)
Supplier:  Cytiva OA001C12
Encompass
Reusable TFF Device for Ultrafiltration Applications. Accelerates and Simplifies Processing of Up to 1 L-Concentration and diafiltration (desalting or buffer exchange) processes can be performed on the same system.-Achieves high concentration factors in a single step due to the low holdup volume of the device, as little as 5 mL.-Capsule includes all fittings and tubing to simplify installation into filtration setups.-Several Minimate TFF capsules may be connected in parallel for increased membrane area. -Capsules have the same path length and materials of construction as larger Pall Centramate™ systems used in process development and production.-Optimized flow path design coupled with low protein binding Omega membrane minimizes non-specific binding.-100% integrity tested during manufacture, for critical applications, the user can re-test the integrity after initial use. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-197-9561