Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPSO, 0.2M buffer soln., pH 9.0, Thermo Scientific™

Catalog No. AAJ61126AK
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61126AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61126AK Supplier Thermo Scientific Chemicals Supplier No. J61126AK

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 102601-34-3
Concentration 0.2 M
pH 9.0
Physical Form Liquid
Quantity 250 mL
Molecular Formula C9H12NO4S
Odor Odorless
MDL Number MFCD00070063
InChI Key CLEVULOKVPBHCU-VIFPVBQESA-M
SMILES O[C@@H](CNC1=CC=CC=C1)CS([O-])(=O)=O
IUPAC Name (2S)-2-hydroxy-3-(phenylamino)propane-1-sulfonate
Molecular Weight (g/mol) 230.26
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.