Learn More
Bioworld CAPSO, Free Acid, 100 g
Supplier:  Bioworld 403300082
Encompass_Preferred
Also known as 3-[Cyclohexylamino]-2-hydroxy-1-propanesulfonic acidA buffer salt used in the electrotransfer of proteins to nitrocellulose membranesUltra Pure Grade Technical Information:Identification(P/F): PassAppearance: SolidDensity : 1.30 g/cm3Melting Point: 270-274 °CPH Value: 8.9 – 10.3
Catalog No. 50-255-801