Learn More
Bioworld CAPSO, Sodium Salt, 100 g
Supplier:  Bioworld 403300122
Encompass_Preferred
Also known as 3-[Cyclohexylamino]-2-hydroxy-1-propanesulfonic acidUsed in immunoblotting experiments, protein sequencing and identificationUltra Pure Grade Technical InformationIdentification(P/F): PassAppearance: SolidMelting Point: 263-271°CPH Value: 8.9 - 10.3
Catalog No. 50-255-803