Learn More
Sigma Aldrich Fine Chemicals Biosciences CAPSO 99 anhydrous basis t500G
Catalog No. 502903428
Encompass
CAPSO is a useful secondary standard buffer. Second dissociation constant pK2 value of CAPSO has been measured. Useful pH range for CAPSO is 8.9 - 10.3.
Catalog No. 50-290-3428 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C2278500G
CAPSO is a useful secondary standard buffer. Second dissociation constant pK2 value of CAPSO has been measured. Useful pH range for CAPSO is 8.9 - 10.3.