Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals CAPSO, 99%

Catalog No. AC327740250
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327740250 25 g Glass bottle
AC327741000 100 g Glass bottle
AC327740010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC327740250 Supplier Thermo Scientific Chemicals Supplier No. 327740250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 73463-39-5 | C9H19NO4S | 237.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73463-39-5
Molecular Formula C9H19NO4S
Molecular Weight (g/mol) 237.3
InChI Key INEWUCPYEUEQTN-UHFFFAOYSA-N
Synonym capso, 3-cyclohexylamino-2-hydroxy-1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxypropane-1-sulfonic acid, capso free acid, 1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxy, capso compound, 3-cyclohexylamino-2-hydroxy-1-propanesuhicic acid, 3-cyclohexylamino-2-hydroxy-1-propanesulphonic acid, 3-cyclohexylamino-2-hydroxypropanesulfonic acid
PubChem CID 2733480
IUPAC Name 3-(cyclohexylamino)-2-hydroxypropane-1-sulfonic acid
SMILES C1CCC(CC1)NCC(CS(=O)(=O)O)O

Specifications

Absorbance (0.1M aq. soln.), 0.03 max. at 280nm, (0.1 M aq. soln.), (0.1 M aq. soln.) at 0nm
Melting Point 263.0°C to 271.0°C
Color White
pH 5 to 6.5 (1% aq. soln. at 25°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Solubility Information (0.1M in water) Clear colorless solution
Formula Weight 237.3
Percent Purity 99%
Physical Form Powder
Chemical Name or Material CAPSO
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.