Learn More
Electron Microscopy Sciences Vacule Caps-Open Top, No Liner,15-425
SDP
Catalog No. 501898930
Encompass_Preferred
Heavy wall construction, designed for lyophilization and freeze-drying. Manufactured from Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class and USP Type I Standards). Accessory for 4 ml Vacule Vials.
Catalog No. 50-189-8930 Supplier Electron Microscopy Sciences Supplier No. 6030443
Heavy wall construction, designed for lyophilization and freeze-drying. Manufactured from Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class and USP Type I Standards). Accessory for 4 ml Vacule Vials.