Learn More
Pyramex™ Capstone™ 500 Series Safety Eyewear
Catalog No. 19155353 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Chemical Green with Neoprene Strap
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color
19-155-353 Chemical Green with Neoprene Strap
19-155-355 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-155-353 Supplier Pyramex™ Supplier No. GC504TN
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fits over prescription glasses
  • Lens is coated to prevent fogging
  • Soft vinyl goggle body conforms to facial contours
  • Goggle provides protection against dust and chemical splash
  • Removable vent caps allow the user to remove when more ventilation is required or leave in place for unsurpassed chemical splash resistance
  • Passes Ballistic MIL-PRF-32432 High Velocity Impact standards
  • Specifications
    Lens Coating Anti-fog
    Lens Type H2X
    Certifications/Compliance ANSI Z87.1
    Lens Tint Clear
    Product Type Goggles
    Frame Color Chemical Green with Neoprene Strap
    Lens Material Polycarbonate