Learn More
Pyramex™ Capstone™ 600 Series Safety Eyewear
Catalog No. 19155320 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint: Clear
Catalog No.
19-155-320 Lens Tint: Clear
1 options
This item is not returnable. View return policy
Catalog No. 19-155-320 Supplier Pyramex™ Supplier No. G604T2
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fits over prescription glasses
  • Indirect ventilation system prevents fogging
  • Includes interchangeable cloth and neoprene straps
  • Co-material goggle provides superior protection against dust entry and chemical splash
  • Thermoplastic (TPE) body with universal nose bridge is flexible and contours to the user's face
  • Specifications
    Lens Coating Anti-fog
    Lens Type H2X
    Certifications/Compliance ANSI Z87.1,CSA Z94.3-07
    Lens Tint Clear
    Frame Color Gray
    Includes Adjustable strap
    Lens Material Polycarbonate