Learn More
Pyramex Safety Products Capstone Goggle
Catalog No. 19555036 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Catalog No. Lens Coating
19-555-036 Anti-fog
1 options
This item is not returnable. View return policy
Catalog No. 19-555-036 Supplier Pyramex Safety Products Supplier No. GG504TM
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex Safety™ Capstone Goggle is a dual purpose goggle adaptable to many different applications.
  • Direct/Indirect-Gray Frame/Clear H2MAX Anti-Fog Lens
  • Removable vent caps allow the user to remove when more ventilation is required or leave in place for chemical resistance
  • Cylinder lens is coated to prevent fogging
  • Soft PVC body conforms to facial contours for comfort fit
  • Fits over prescription glasses
Specifications
Lens Coating Anti-fog
Lens Tint Clear
Frame Color Gray
Frame Material PVC