Learn More
Braintree Scientific Capsule Dosing Syringe, Stainless Steel, for size 9el
SDP
Catalog No. 501956160
Encompass
Stainless Steel Capsule Dosing SyringeIf dosing multiple animals, order additional syringes to allow them to dry between administrations. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6160 Supplier Braintree Scientific Supplier No. XSYR9EL
Stainless Steel Capsule Dosing SyringeIf dosing multiple animals, order additional syringes to allow them to dry between administrations. **Sold for research purposes only and not for human use**