Learn More
Braintree Scientific Capsule Dosing Syringe,Stainless Steel, for size 9H kit
SDP
Supplier:  Braintree Scientific XSYR9H
Encompass
Mini Capsules Dosing SyringeIf dosing multiple animals, order additional syringes to allow them to dry between administrations. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6161