Learn More
Electron Microscopy Sciences Capsule Mold Type C, 12 Cavities
SDP
Supplier:  Electron Microscopy Sciences 70161
Encompass_Preferred
Our molds are made from the strongest possible silicone rubber providing extremely high tear resistance. The molds are are reusable. Type C has 12 cavities cylindrical shapes, 8mm diameter 16mm height, flat tip.
Catalog No. 50-248-66