Learn More
Electron Microscopy Sciences mPrep/g™ Capsules with Labels in storage box
SDP
Supplier:  Electron Microscopy Sciences 8501004
Encompass_Preferred
Each capsule can hold one or two TEM grids securely for staining, immuno-labeling and labeled storage. Grids are protected from loss, misidentification, and damage. Grid-tite slots keep grids safe even if an open capsule is dropped.
Grids require handling only twice: when inserted into capsules and when placed in the TEM.
Using a multi-channel pipettor, processing up to 24 grids simultaneously takes no more effort than a single grid. Reagent consumption is as little as 20 uL per grid. The chance of grid damage or loss is greatly reduced using these capsules.
Catalog No. 50-192-8114