Learn More
Cytiva (Formerly Pall Lab) Mini Profile Capsules - Profile Star Filter, 1.5 m (3/pkg)
Supplier:  Cytiva 12070
Encompass
Scaleable Depth Capsules for Prefiltration and Clarification-High capacity depth medium is absolute particle rated for reliability.-Filter materials offer broad chemical compatibility.-Optimized for viscous fluids. -Mini Profile II filters have a thick depth structure, providing high capacity for larger solids and gels as well as for fine particles.-Profile Star filters feature high area star-shaped pleated construction. The patented design combines the advantages of thick depth filters with those of traditional high area pleated filters. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3173