Learn More
Braintree Scientific Capsule Sinker, for size 9 capsules
SDP
Catalog No. 501956155
Encompass
CapsuleSinker Allows you to determine the dissolution time for a single capsule. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6155 Supplier Braintree Scientific Supplier No. XSINK9
CapsuleSinker Allows you to determine the dissolution time for a single capsule. **Sold for research purposes only and not for human use**