Learn More
Braintree Scientific Capsule Sinker, for size M capsules
SDP
Catalog No. 501956158
Encompass
MultipleCapsule Filler Fill 40 capsules at a time to withinin +/- 7-10% of average weight Includes filler, tamper, and spreader **Sold for research purposes only and not for human use**
Catalog No. 50-195-6158 Supplier Braintree Scientific Supplier No. XSINKM
MultipleCapsule Filler Fill 40 capsules at a time to withinin +/- 7-10% of average weight Includes filler, tamper, and spreader **Sold for research purposes only and not for human use**