Learn More
Quality Lab Accessories Spiral Capsule Sinker, 316 Stainless Steel, Electropolished Finish, 1.15 in Length
Catalog No. NC0878446
Encompass
The Spiral Capsule Sinker is engineered for dissolution testing, ensuring effective stabilization of floating dosage forms for reproducible results. Made from durable electropolished 316 Stainless Steel, it accommodates capsule sizes 2 to 000 and is compatible with various dissolution baths. This high-quality sinker enhances testing reliability, making it a critical tool in pharmaceutical analysis.
Catalog No. NC0878446 Supplier Quality Lab Accessories Supplier No. CAPWHTXLS
The Spiral Capsule Sinker is engineered for dissolution testing, ensuring effective stabilization of floating dosage forms for reproducible results. Made from durable electropolished 316 Stainless Steel, it accommodates capsule sizes 2 to 000 and is compatible with various dissolution baths. This high-quality sinker enhances testing reliability, making it a critical tool in pharmaceutical analysis.