Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Capsule Stain Kit (Hiss Method)

Catalog No. ES6890
Change view
Click to view available options
Quantity:
2 x 100 mL
2 x 250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6890 2 x 250 mL
ES6880 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. ES6890 Supplier ENG Scientific Supplier No. 6890
Add to Cart
Add to Cart
This item is not returnable. View return policy

With the Capsule Stain Kit (Hiss Method), capsules appear as faint Blue halos around blue to purple cells. Includes Solution I and Solution II.

  • Capsule Stain Kit
  • Contents: Crystal Violet, Copper Sulfate
  • Solution I - Aqueous Crystal Violet (1%) 250 mL and Solution II - Aqueous Copper Sulfate (20%) 250 mL
  • For in vitro diagnostic use
  • 1 Kit
  • Solution made from certified dyes (when applicable)
  • Recommended Procedure:
    • Grow organisms in ascitic fluid or serum medium, or mix a drop of serum with specimen and from this mixture, prepare smear.
    • Air dry.
    • Cover smear with Sol. for 2 minutes.
    • Wash off the stain with Sol. II.
    • The amount of washing must be determined empirically.
  • Results: Capsules appear as faint blue halos around dark blue to purple cells.

Specifications

Quantity 2 x 250 mL
Includes Solution I - Aqueous Crystal Violet (1%), Solution II - Aqueous Copper Sulfate (20%)
For Use With (Application) In vitro diagnostic use
Volume (Metric) 250 mL
Product Type Capsule Stain Kit
Packaging Bottles

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.