Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Capsule Stain Kit (Hiss Method)

Catalog No. p-7226650
Change view
Click to view available options
Quantity:
2 x 100 mL
2 x 250 mL
Catalog No. Quantity
ES6880 2 x 100 mL
ES6890 2 x 250 mL
2 options

Catalog No. ES6880

Supplier: ENG Scientific 6880

Only null left
Add to Cart
Add to Cart

With the Capsule Stain Kit (Hiss Method), capsules appear as faint Blue halos around blue to purple cells. Includes Solution I and Solution II.

  • Capsule Stain Kit
  • Contents: Crystal Violet, Copper Sulfate
  • Solution I - Aqueous Crystal Violet (1%) 250 mL and Solution II - Aqueous Copper Sulfate (20%) 250 mL
  • For in vitro diagnostic use
  • 1 Kit
  • Solution made from certified dyes (when applicable)
  • Recommended Procedure:
    • Grow organisms in ascitic fluid or serum medium, or mix a drop of serum with specimen and from this mixture, prepare smear.
    • Air dry.
    • Cover smear with Sol. for 2 minutes.
    • Wash off the stain with Sol. II.
    • The amount of washing must be determined empirically.
  • Results: Capsules appear as faint blue halos around dark blue to purple cells.

Specifications

Type Capsule Stain Kit
Quantity 2 x 100 mL
Includes Solution I - Aqueous Crystal Violet (1%), Solution II - Aqueous Copper Sulfate (20%)
Packaging Type Bottles

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.