Learn More
Labelmaster Inc CAPSULOC™ SPECIAL PERMIT ASSEMBLED KIT, UP TO 32 OZ. BOTTLE
SDP
Catalog No. NC2587397
Encompass_Preferred
Help safely ship your hazardous materials using your own bottles with this 4GV/4G kit featuring the innovative Capsuloc™ secondary container. Capsuloc has been issued Special Permit DOT-SP 21488 and provides a lighter, safer, and more convenient solution for hazmat containment compared to traditional metal paint can secondary containers.
Catalog No. NC2587397 Supplier Labelmaster Inc Supplier No. UNIPCAPSULOCSP32
Help safely ship your hazardous materials using your own bottles with this 4GV/4G kit featuring the innovative Capsuloc™ secondary container. Capsuloc has been issued Special Permit DOT-SP 21488 and provides a lighter, safer, and more convenient solution for hazmat containment compared to traditional metal paint can secondary containers.