Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ CapSure™ LCM MicroCaps

Catalog No. 501148720
Encompass_Preferred
Change view
Click to view available options
Quantity:
12 Preps
48 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-114-8720 48 Preps
50-114-8721 12 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-114-8720 Supplier Applied Biosystems™ Supplier No. A30153
Add to Cart
Edge
Add to Cart

Specially designed for use with the Arcturus™ Laser Capture Microdissection (LCM) instruments, CapSure™ Micro LCM Caps enable the precise and rapid extraction of pure populations of cells from heterogeneous sample preparations.

CapSure Micro LCM Caps may be used to isolate individual cells or several hundreds of cells on a single cap.

Key product features:
Gentle application—non-damaging microdissection preserves the integrity of the captured material
Versatile extraction—capture both large and small amounts of cells

Supports the integrity of microdissected cells for downstream molecular analysis
All CapSure LCM Caps have a proprietary transfer film bonded to the lower cap surface. When using the Arcturus microdissection systems an infrared laser pulses through the top of the cap during LCM and interacts with the transfer film, which then melts and bonds to the cells or regions of interest. The film absorbs the laser radiation—instead of the tissue or cell sample—creating a gentle, non-damaging microdissection that preserves the integrity of the captured material.

Go straight from capture to downstream analysis
CapSure Micro LCM Caps enable genetic material extraction with as little as 10 μL of lysis buffer. The new and improved design now enables the use of 0.2 mL microcentrifuge tubes. With this unique combination, you can proceed directly from LCM to downstream applications such as qPCR and next-generation sequencing with a single-tube workflow, while preserving precious sample.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 48 Preps
Product Type LCM Cap
Content And Storage • 48 CapSure LCM MicroCaps
• 96 Dome caps
For Use With (Equipment) ArcturusXT™ LCM Instrument
Shipping Condition Room Temperature
Product Line Arcturus, CapSure

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.