Learn More
Sigma Aldrich Fine Chemicals Biosciences Capto Adhere ImpRes 100 ML
Catalog No. 502907249
Encompass
The multimodal anion exchanger ligand of Capto adhere ImpRes is immobilized to the base matrix and interacts with the target molecule through multiple types of interactions.The high selectivity of the anion multimodal ligand allows operation in a variety of process conditions to solve challenging purification problems. Contaminants such as DNA host cell proteins leached protein A aggregates and viruses are efficiently removed.Capto adhere ImpRes is available in a range of different bulk pack sizes for easy scale-up and pre-packed formats for process development.As member of the BioProcess media range Capto adhere ImpRes meets industrial demands with security of supply and comprehensive technical and regulatory support.
Catalog No. 50-290-7249 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17371502
The multimodal anion exchanger ligand of Capto adhere ImpRes is immobilized to the base matrix and interacts with the target molecule through multiple types of interactions.The high selectivity of the anion multimodal ligand allows operation in a variety of process conditions to solve challenging purification problems. Contaminants such as DNA host cell proteins leached protein A aggregates and viruses are efficiently removed.Capto adhere ImpRes is available in a range of different bulk pack sizes for easy scale-up and pre-packed formats for process development.As member of the BioProcess media range Capto adhere ImpRes meets industrial demands with security of supply and comprehensive technical and regulatory support.