Learn More
Sigma Aldrich Fine Chemicals Biosciences Capto MMC ImpRes 100 ML Cy
Catalog No. 502907107
Encompass
The weak cation exchange multimodal ligand enables high selectivity in a broad pH/salt window compared with traditional ion exchangers which allows the use of Capto MMC ImpRes in a variety of process conditions to solve challenging purification problems.
Catalog No. 50-290-7107 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17371602
The weak cation exchange multimodal ligand enables high selectivity in a broad pH/salt window compared with traditional ion exchangers which allows the use of Capto MMC ImpRes in a variety of process conditions to solve challenging purification problems.