Learn More
Apexbio Technology LLC Captopril, 100mg. Cas: 62571-86-2 MFCD: MFCD00168073. ACE inhibitor
SDP
Supplier:  Apexbio Technology LLC A4078100
Encompass_Preferred
Captopril is an inhibitor of Angiotensin-I-converting enzyme (ACE) with IC50 value of 6nM [1].Captopril is an old drug with antihypertensive efficacy commonly used in the control of blood pressure. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4873