Learn More
Enzo Life Sciences CaptoprIL-(1g). CAS: 62571-86-2
SDP
Supplier:  Enzo Life Sciences ALX270212G001
Encompass_Preferred
First orally active angiotensin-converting enzyme (ACE) inhibitor. IC50=2.14nM. Reversible and competitive inhibitor of LTA4 hydrolase (IC50=11μM). Inhibits angiogenesis. Antihypotensive agent. Alternative name: (2S)-1-(3-Mercapto-2-methylpropionyl)-L-proline. Formula: C9H15NO3S. MW: 217.3. Purity: ≥98% (HPLC). Solubility: Soluble in water, chloroform, dichloromethane or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8429