Learn More
Abcam Captopril, ACE inhibitor, 1G
Catalog No. 502706672
Encompass_Preferred
MW 217.29 Da, Purity >98%. Potent ACE inhibitor (IC₅₀ = 21 nM). Reversible and competitive leukotriene A₄ hydrolase inhibitor (Ki = 6 μM, leukotriene B4 formation). Vasodilatory and antihypertensive effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6672 Supplier Abcam Supplier No. AB1413331G
MW 217.29 Da, Purity >98%. Potent ACE inhibitor (IC₅₀ = 21 nM). Reversible and competitive leukotriene A₄ hydrolase inhibitor (Ki = 6 μM, leukotriene B4 formation). Vasodilatory and antihypertensive effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement