Learn More
Cayman Chemical CaptoprIl-d3 100ug
SDP
Supplier:  Cayman Chemical 25235100 UG
Encompass_Preferred
An internal standard for the quantification of captopril by GC- or LC-MS
Catalog No. 50-293-2631