Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Captopril-d3 | 1356383-38-4 | 100 UG
SDP

Catalog No. 5000406615
Encompass_Preferred

Captopril-d3 is deuterium labeled Captopril. It is an antihypertensive agent and a thiol-containing competitive, orally active angiotensin-converting enzyme (ACE) inhibitor (IC50=0.025 μM). It has been widely used in research for hypertension and congestive heart failure. Captopril-d3 also acts as a New Delhi metallo-β-lactamase-1 (NDM-1) inhibitor with an IC50 of 7.9 μM. It is for research use only.

  • Can be used as a tracer for drug development
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration affects pharmacokinetic and metabolic profiles of drugs
  • Molecular weight: 220.30
  • Chemical formula: C9H12D3NO3S

Catalog No. 50-004-06615 Supplier Medchemexpress LLC Supplier No. HYB0368S100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Captopril-d3 is deuterium labeled Captopril. It is an antihypertensive agent and a thiol-containing competitive, orally active angiotensin-converting enzyme (ACE) inhibitor (IC50=0.025 μM). It has been widely used in research for hypertension and congestive heart failure. Captopril-d3 also acts as a New Delhi metallo-β-lactamase-1 (NDM-1) inhibitor with an IC50 of 7.9 μM. It is for research use only.

  • Can be used as a tracer for drug development
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration affects pharmacokinetic and metabolic profiles of drugs
  • Molecular weight: 220.30
  • Chemical formula: C9H12D3NO3S

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.